2-Iodopyridine, 95%
2-Iodopyridine, 95%
2-Iodopyridine, 95%
2-Iodopyridine, 95%
Thermo Scientific Chemicals

2-Iodopyridine, 95%

CAS: 5029-67-4 | C5H4IN | 205.00 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 H63062.14
亦稱為 H63062-14
價格 (TWD)
-
申請報價
Quantity:
25 g
Request bulk or custom format
化學識別
CAS5029-67-4
規格Specification Sheet規格表
Assay (HPLC)>94.0%
FormLow melting solid
Appearance (Color)Colorless to yellow
2-Iodopyridine is a reagent used in the preparation of human NAD+-dependent 15-hydroxyprostaglandin dehydrogenase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodopyridine is a reagent used in the preparation of human NAD+-dependent 15-hydroxyprostaglandin dehydrogenase inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Damien Y. Duveau; Adam Yasgar; Yuhong Wang; Xin Hu; Jennifer Kouznetsova; Kyle R. Brimacombe; Ajit Jadhav; Anton Simeono; Craig J. Thomas; David J. Maloneyl. Structure-activity relationship studies and biological characterization of human NAD+-dependent 15-hydroxyprostaglandin dehydrogenase inhibitors. Bioorganic & Medicinal Chemistry Letters. 2014, 24 (2), 401-409.
  2. H- H. Tai; H. Cho, M. Tong; and Y. Ding. NAD+-Linked 15-Hydroxyprostaglandin Dehydrogenase: Structure and Biological Functions. Curr. Pharm. Des. 2006, 12 (8), 955 - 962.