Methyl 3-cyanoindole-6-carboxylate, 97%
Methyl 3-cyanoindole-6-carboxylate, 97%
Methyl 3-cyanoindole-6-carboxylate, 97%
Methyl 3-cyanoindole-6-carboxylate, 97%
Thermo Scientific Chemicals

Methyl 3-cyanoindole-6-carboxylate, 97%

CAS: 1000576-51-1 | C11H8N2O2 | 200.197 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H63100.MD
also known as H63100-MD
Price (EUR)
73,30
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
73,30
Each
Add to cart
Specifications
CAS1000576-51-1
Chemical Name or MaterialMethyl 3-cyanoindole-6-carboxylate
Formula Weight200.19
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only