Thermo Scientific Chemicals

D-Tyrosine tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H63274.06
5 g, Each
Thermo Scientific Chemicals

D-Tyrosine tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H63274.06
5 g, Each
Quantity
Catalog number: H63274.06
also known as H63274-06
Price (USD)
Price: 122.00
Your price:
Quantity
-

Chemical Identifiers

CAS
87553-74-0
IUPAC Name
tert-butyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate
Molecular Formula
C13H19NO3
InChI Key
DIGHFXIWRPMGSA-LDGXTIHJNA-N
SMILES
CC(C)(C)OC(=O)[C@H](N)CC1=CC=C(O)C=C1
Assay (HPLC)
>97.5%

Description

D-Tyrosine tert-butyl ester, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Tyrosine tert-butyl ester, is used as a pharmaceutical intermediate.

Solubility
Very slightly soluble in cold water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text