1-Benzylindole, 97%
1-Benzylindole, 97%
1-Benzylindole, 97%
Thermo Scientific Chemicals

1-Benzylindole, 97%

CAS: 3377-71-7 | C15H13N | 207.276 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63414.03
also known as H63414-03
Price (TWD)
-
Request A Quote
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein148884
CAS3377-71-7
Chemical Name or Material1-Benzylindole
ColorWhite to Yellow
Formula Weight207.27
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only