Thermo Scientific Chemicals

N-Fmoc-D-allo-isoleucine, 98%, Thermo Scientific Chemicals

Catalog number: H63455.06
5 g, Each
Thermo Scientific Chemicals

N-Fmoc-D-allo-isoleucine, 98%, Thermo Scientific Chemicals

Catalog number: H63455.06
5 g, Each
Quantity
Catalog number: H63455.06
also known as H63455-06
Price (USD)
Price: 860.00
Online price: 731.65
Your price:
Quantity
-

Chemical Identifiers

CAS
118904-37-3
IUPAC Name
(2R,3S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylpentanoic acid
Molecular Formula
C21H23NO4
InChI Key
QXVFEIPAZSXRGM-KINVLMLONA-N
SMILES
CC[C@H](C)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Assay (HPLC)
>97.5%

Description

N-Fmoc-D-allo-isoleucine used as a intermediate in pharmaceutical and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-D-allo-isoleucine used as a intermediate in pharmaceutical and chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate at -20°C. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text