Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine, 95%
Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine, 95%
Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine, 95%
Thermo Scientific Chemicals

Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine, 95%

CAS: 146987-10-2 | C31H38N4O6S | 594.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63609.03
also known as H63609-03
Price (USD)
271.65
Online Exclusive
302.00
Save 30.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
271.65
Online Exclusive
302.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS146987-10-2
IUPAC Name(2S)-6-{5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanamido}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Molecular FormulaC31H38N4O6S
InChI KeyOFIBQNGDYNGUEZ-CVKYCUBCNA-N
SMILES[H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](NC(=O)OCC3C4=CC=CC=C4C4=CC=CC=C34)C(O)=O)[C@@]1([H])NC(=O)N2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%

Nα-Fmoc-Nε-biotinyl-L-lysine is useful in the synthesis of site-specific biotinylated probes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nα-Fmoc-Nε-biotinyl-L-lysine is useful in the synthesis of site-specific biotinylated probes.

Solubility
Slightly soluble in water and dimethyl sulfoxide.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
M J Macielag; T Peeters; I Depoortere. Synthesis and characterization of site-specific biotinylated probes for the motilin receptor. International Journal of Peptide and Protein Reseach. 1994, 44(6), 582-588.