N-Fmoc-O-tert-butyl-N-methyl-L-serine, 97%, Thermo Scientific Chemicals
N-Fmoc-O-tert-butyl-N-methyl-L-serine, 97%, Thermo Scientific Chemicals
N-Fmoc-O-tert-butyl-N-methyl-L-serine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Fmoc-O-tert-butyl-N-methyl-L-serine, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H63614.06
also known as H63614-06
5 g
Catalog number H63614.06
also known as H63614-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS197632-77-2
IUPAC Name3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)propanoic acid
Molecular FormulaC23H27NO5
InChI KeyPQSAXALAXPNFMG-UHFFFAOYNA-N
SMILESCN(C(COC(C)(C)C)C(O)=O)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only