Thermo Scientific Chemicals

Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine, 95%, Thermo Scientific Chemicals

Catalog number: H63629.03
1 g, Each
Thermo Scientific Chemicals

Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine, 95%, Thermo Scientific Chemicals

Catalog number: H63629.03
1 g, Each
Quantity
Catalog number: H63629.03
also known as H63629-03
Price (USD)
Price: 76.70
Online price: 65.65
Your price:
Quantity
-

Chemical Identifiers

CAS
167393-62-6
IUPAC Name
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(4-methylphenyl)diphenylmethyl]amino}hexanoic acid
Molecular Formula
C41H40N2O4
InChI Key
YPTNAIDIXCOZAJ-WHUAGQPENA-N
SMILES
CC1=CC=C(C=C1)C(NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
Assay (HPLC)
>94.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text