2-Bromo-3-fluorobenzyl bromide, 98%
2-Bromo-3-fluorobenzyl bromide, 98%
2-Bromo-3-fluorobenzyl bromide, 98%
2-Bromo-3-fluorobenzyl bromide, 98%
Thermo Scientific Chemicals

2-Bromo-3-fluorobenzyl bromide, 98%

CAS: 1184918-22-6 | C7H5Br2F | 267.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63632.06
also known as H63632-06
Price (TWD)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1184918-22-6
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98.0% min
It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only