Thermo Scientific Chemicals

D-Aspartic acid 1-methyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H63658.14
25 g, Each
Thermo Scientific Chemicals

D-Aspartic acid 1-methyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H63658.14
25 g, Each
Quantity
Catalog number: H63658.14
also known as H63658-14
Price (USD)
Price: 320.00
Online price: 276.65
Your price:
Quantity
-

Chemical Identifiers

CAS
65414-78-0
IUPAC Name
3-amino-4-methoxy-4-oxobutanoic acid
Molecular Formula
C5H9NO4
InChI Key
SWWBMHIMADRNIK-UHFFFAOYNA-N
SMILES
COC(=O)C(N)CC(O)=O
Assay (Titration)
>97.5%

Description

D-Aspartic acid 1-methyl ester is used as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Aspartic acid 1-methyl ester is used as a intermediate for pharmaceutical and chemical research.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text