Thermo Scientific Chemicals

L-Glutamic acid di-tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H63708.06
5 g, Each
Thermo Scientific Chemicals

L-Glutamic acid di-tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H63708.06
5 g, Each
Quantity
Catalog number: H63708.06
also known as H63708-06
Price (USD)
Price: 143.00
Online price: 121.65
Your price:
Quantity
-

Chemical Identifiers

CAS
32677-01-3
IUPAC Name
1,5-di-tert-butyl 2-aminopentanedioate hydrochloride
Molecular Formula
C13H26ClNO4
InChI Key
LFEYMWCCUAOUKZ-UHFFFAOYNA-N
SMILES
Cl.CC(C)(C)OC(=O)CCC(N)C(=O)OC(C)(C)C
Assay from Suppliers CofA
≥97.5% (HPLC)
Appearance (Color)
White to pale cream or pale yellow
Form
Powder or crystals or crystalline powder
Identification (FTIR)
Conforms to structure
Water Content (Karl Fischer Titration)
≤0.5%

Description

L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.

Solubility
Soluble in methanol (50gmg/ml).

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text