trans-N-Fmoc-4-tert-butoxy-L-proline, 98%
trans-N-Fmoc-4-tert-butoxy-L-proline, 98%
trans-N-Fmoc-4-tert-butoxy-L-proline, 98%
trans-N-Fmoc-4-tert-butoxy-L-proline, 98%
Thermo Scientific Chemicals

trans-N-Fmoc-4-tert-butoxy-L-proline, 98%

CAS: 122996-47-8 | C24H27NO5 | 409.482 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63791.03
also known as H63791-03
Price (EUR)
62,90
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
62,90
Each
Add to cart
Chemical Identifiers
CAS122996-47-8
IUPAC Name4-(tert-butoxy)-1-{[(9H-fluoren-9-yl)methoxy]carbonyl}pyrrolidine-2-carboxylic acid
Molecular FormulaC24H27NO5
InChI KeyWPBXBYOKQUEIDW-UHFFFAOYNA-N
SMILESCC(C)(C)OC1CC(N(C1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only