3-(1H-Tetrazol-1-yl)aniline, 97%
3-(1H-Tetrazol-1-yl)aniline, 97%
3-(1H-Tetrazol-1-yl)aniline, 97%
Thermo Scientific Chemicals

3-(1H-Tetrazol-1-yl)aniline, 97%

CAS: 14213-12-8 | C7H7N5 | 161.168 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H63829.03
also known as H63829-03
Price (EUR)
141,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
141,00
Each
Specifications
CAS14213-12-8
Chemical Name or Material3-(1H-Tetrazol-1-yl)aniline
Formula Weight161.16
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only