4-(2-Fluorobenzyl)piperidine hydrochloride, 97%
4-(2-Fluorobenzyl)piperidine hydrochloride, 97%
4-(2-Fluorobenzyl)piperidine hydrochloride, 97%
Thermo Scientific Chemicals

4-(2-Fluorobenzyl)piperidine hydrochloride, 97%

CAS: 193357-21-0 | C12H17ClFN | 229.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63849.06
also known as H63849-06
Price (EUR)
775,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
775,00
Each
Specifications
CAS193357-21-0
Chemical Name or Material4-(2-Fluorobenzyl)piperidine hydrochloride
ColorWhite
Formula Weight229.72
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only