N-Fmoc-3-cyclohexyl-L-alanine, 98%
N-Fmoc-3-cyclohexyl-L-alanine, 98%
N-Fmoc-3-cyclohexyl-L-alanine, 98%
Thermo Scientific Chemicals

N-Fmoc-3-cyclohexyl-L-alanine, 98%

CAS: 135673-97-1 | C24H27NO4 | 393.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H63910.03
also known as H63910-03
Price (EUR)
66,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
66,60
Each
Chemical Identifiers
CAS135673-97-1
IUPAC Name3-cyclohexyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Molecular FormulaC24H27NO4
InChI KeyHIJAUEZBPWTKIV-UHFFFAOYNA-N
SMILESOC(=O)C(CC1CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystalline powder or powder or solid
Assay≥96.0%
Assay (unspecified)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only