1-Fmoc-piperidine-4-carboxylic acid, 98%
1-Fmoc-piperidine-4-carboxylic acid, 98%
1-Fmoc-piperidine-4-carboxylic acid, 98%
Thermo Scientific Chemicals

1-Fmoc-piperidine-4-carboxylic acid, 98%

CAS: 148928-15-8 | C21H21NO4 | 351.402 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H63915.14
also known as H63915-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS148928-15-8
IUPAC Name1-{[(9H-fluoren-9-yl)methoxy]carbonyl}piperidine-4-carboxylic acid
Molecular FormulaC21H21NO4
InChI KeyOJYOOHSQOIDDOO-UHFFFAOYSA-N
SMILESOC(=O)C1CCN(CC1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only