3-(Dimethylaminomethyl)-5-nitroindole, 95%, Thermo Scientific Chemicals
3-(Dimethylaminomethyl)-5-nitroindole, 95%, Thermo Scientific Chemicals
3-(Dimethylaminomethyl)-5-nitroindole, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(Dimethylaminomethyl)-5-nitroindole, 95%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H63939.03
also known as H63939-03
1 g
Catalog number H63939.03
also known as H63939-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein216692
CAS3414-64-0
Chemical Name or Material3-(Dimethylaminomethyl)-5-nitroindole
Formula Weight219.24
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only