1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride, 97%
1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride, 97%
1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride, 97%
Thermo Scientific Chemicals

1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride, 97%

CAS: 346451-15-8 | C30H30Cl4N2 | 560.384 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H63961.06
also known as H63961-06
Price (EUR)
366,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
366,00
Each
Specifications
CAS346451-15-8
Chemical Name or Material1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride
ColorWhite
Formula Weight560.38
IUPAC Name1,4-bis[(4-chlorophenyl)-phenylmethyl]piperazine;dihydrochloride
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only