5-Cyano-2-fluoropyridine, 95%
5-Cyano-2-fluoropyridine, 95%
Thermo Scientific Chemicals

5-Cyano-2-fluoropyridine, 95%

CAS: 3939-12-6 | C7H8BrN | 186.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64278.03
also known as H64278-03
Price (EUR)
76,40
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
76,40
Each
Add to cart
Chemical Identifiers
CAS3939-12-6
IUPAC Name6-fluoropyridine-3-carbonitrile
Molecular FormulaC6H3FN2
InChI KeyVRLVOMUVHHHJHB-UHFFFAOYSA-N
SMILESFC1=NC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only