4-Bromo-2,6-difluorobenzotrifluoride, 98%
4-Bromo-2,6-difluorobenzotrifluoride, 98%
4-Bromo-2,6-difluorobenzotrifluoride, 98%
Thermo Scientific Chemicals

4-Bromo-2,6-difluorobenzotrifluoride, 98%

CAS: 156243-64-0 | C7H2BrF5 | 260.989 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 H64386.03
亦稱為 H64386-03
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
化學識別
CAS156243-64-0
IUPAC Name5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene
Molecular FormulaC7H2BrF5
InChI KeyQPJKIRNIIXIPIE-UHFFFAOYSA-N
SMILESFC1=CC(Br)=CC(F)=C1C(F)(F)F
檢視更多
規格Specification Sheet規格表
Assay (GC)>97.5%
It is employed in the lewis acid-promoted synthesis of unsymmetrical and highly functionalized carbazoles and dibenzofurans from biaryl triazenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the lewis acid-promoted synthesis of unsymmetrical and highly functionalized carbazoles and dibenzofurans from biaryl triazenes.

Solubility
Slightly soluble in water.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat.
RUO – Research Use Only
Weijun Yang, et al. Lewis Acid-Promoted Synthesis of Unsymmetrical and Highly Functionalized Carbazoles and Dibenzofurans from Biaryl Triazenes: Application for the Total Synthesis of Clausine C, Clausine R, and Clauraila A.Chem. Eur. J.,2011,17(49), 13665-13669.