Thioisobutyramide, 95%
Thioisobutyramide, 95%
Thioisobutyramide, 95%
Thermo Scientific Chemicals

Thioisobutyramide, 95%

CAS: 13515-65-6 | C4H9NS | 103.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H64663.06
also known as H64663-06
Price (TWD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS13515-65-6
IUPAC Name2-methylpropanethioamide
Molecular FormulaC4H9NS
InChI KeyNPCLRBQYESMUPD-UHFFFAOYSA-N
SMILESCC(C)C(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only