5-Bromo-2-hydroxy-4-methylpyridine, 97%
5-Bromo-2-hydroxy-4-methylpyridine, 97%
5-Bromo-2-hydroxy-4-methylpyridine, 97%
Thermo Scientific Chemicals

5-Bromo-2-hydroxy-4-methylpyridine, 97%

CAS: 164513-38-6 | C6H6BrNO | 188.024 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64674.06
also known as H64674-06
Price (EUR)
55,40
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
55,40
Each
Chemical Identifiers
CAS164513-38-6
IUPAC Name5-bromo-4-methyl-1,2-dihydropyridin-2-one
Molecular FormulaC6H6BrNO
InChI KeyBYGNXKDNIFHZHQ-UHFFFAOYSA-N
SMILESCC1=CC(=O)NC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
It is used for medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for medicine.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only