4-(Trifluoromethyl)imidazole, 98%
4-(Trifluoromethyl)imidazole, 98%
4-(Trifluoromethyl)imidazole, 98%
4-(Trifluoromethyl)imidazole, 98%
Thermo Scientific Chemicals

4-(Trifluoromethyl)imidazole, 98%

CAS: 33468-69-8 | C4H3F3N2 | 136.077 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64711.03
also known as H64711-03
Price (EUR)
41,00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
41,00
Each
Add to cart
Chemical Identifiers
CAS33468-69-8
IUPAC Name5-(trifluoromethyl)-1H-imidazole
Molecular FormulaC4H3F3N2
InChI KeyDFLGRTIPTPCKPJ-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CN=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only