1-Bromo-4-iodonaphthalene, 98%
1-Bromo-4-iodonaphthalene, 98%
1-Bromo-4-iodonaphthalene, 98%
Thermo Scientific Chemicals

1-Bromo-4-iodonaphthalene, 98%

CAS: 63279-58-3 | C10H6BrI | 332.966 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64825.03
also known as H64825-03
Price (EUR)
73,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
73,30
Each
Chemical Identifiers
CAS63279-58-3
IUPAC Name1-bromo-4-iodonaphthalene
Molecular FormulaC10H6BrI
InChI KeyHQHHKYXPFKHLBF-UHFFFAOYSA-N
SMILESBrC1=CC=C(I)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
It is used as an OLED material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an OLED material.

Solubility
Slightly soluble in water. (3.6 g/L) (25°C)

Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Chris Petucci.; Marianne Nyman.; Leo Guler.; Hilkka Kenttäma. Hydrogen Atom Abstraction Reactions of Charged Polyaromatic σ-Radicals Related to the Active Intermediates of the Enediyne Antitumor Drugs.J. Am. Chem. Soc. 2002, 124 (15), 4108-4115 .
  2. Linhong Jing.; John J. Nash.; Hilkka I. Kenttäma. Correlation of Hydrogen-Atom Abstraction Reaction Efficiencies for Aryl Radicals with their Vertical Electron Affinities and the Vertical Ionization Energies of the Hydrogen-Atom Donors.J. Am. Chem. Soc. 2008, 130 (52),17697-17709 .