4-(Diphenylamino)benzaldehyde, 98%
4-(Diphenylamino)benzaldehyde, 98%
4-(Diphenylamino)benzaldehyde, 98%
4-(Diphenylamino)benzaldehyde, 98%
Thermo Scientific Chemicals

4-(Diphenylamino)benzaldehyde, 98%

CAS: 4181-05-9 | C19H15NO | 273.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H64887.14
also known as H64887-14
Price (EUR)
493,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
493,00
Each
Add to cart
Chemical Identifiers
CAS4181-05-9
IUPAC Name4-(diphenylamino)benzaldehyde
Molecular FormulaC19H15NO
InChI KeyUESSERYYFWCTBU-UHFFFAOYSA-N
SMILESO=CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow
FormPowder or crystals or crystalline powder
Infrared spectrumConforms
Purity≥97.5%
It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only
Hiroshi Inada.; Yasuhiko Shirota. 1,3,5-Tris[4-(diphenylamino)phenyl]benzene and its methyl-substituted derivatives as a novel class of amorphous molecular materials. J. Mater. Chem. 1993, 3 (3), 319-320 .