Dibenzofuran-2-carboxaldehído, 98 %, Thermo Scientific Chemicals
Dibenzofuran-2-carboxaldehído, 98 %, Thermo Scientific Chemicals
Dibenzofuran-2-carboxaldehído, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dibenzofuran-2-carboxaldehído, 98 %, Thermo Scientific Chemicals

CAS: 5397-82-0 | C13H8O2 | 196.21 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo H64905.06
también denominado H64905-06
Precio (USD)
-
Cantidad:
5 g
Pedido a granel o personalizado
Identificadores químicos
CAS5397-82-0
IUPAC Name8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene-4-carbaldehyde
Molecular FormulaC13H8O2
InChI KeyOVJMIWIVPWPZMN-UHFFFAOYSA-N
SMILESO=CC1=CC2=C(OC3=C2C=CC=C3)C=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Assay from Supplier's CofA≥97.5%
Appearance (Color)Pale cream to pale yellow
FormPowder
Used as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
Se utiliza como intermedio farmacéutico.

Notas
Sensible al aire. Almacenar alejada del aire. Almacenar en un lugar fresco y seco en un recipiente cerrado herméticamente. Garantice una ventilación adecuada. Almacenar alejado de agentes oxidantes.
RUO - Para uso exclusivo en investigación
  1. Thirumal Yempala; Jonnalagadda Padma Sridevi; Perumal Yogeeswari; Dharmarajan Sriram; Srinivas Kantevari. Rational design and synthesis of novel dibenzo[b,d]furan-1,2,3-triazole conjugates as potent inhibitors of Mycobacterium tuberculosis. European Journal of Medicinal Chemistry.2014 7(71), 160-167.