Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-leucine N-succinimidyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H65546.22
100 g, Each
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-leucine N-succinimidyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H65546.22
100 g, Each
Quantity
Catalog number: H65546.22
also known as H65546-22
Price (USD)
Price: 655.00
Online price: 566.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3397-35-1
IUPAC Name
2,5-dioxopyrrolidin-1-yl (2S)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoate
Molecular Formula
C18H22N2O6
InChI Key
YHZUOMRURVTBMO-YQTOOIBONA-N
SMILES
CC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
Appearance (Color)
White
Assay (HPLC)
≥94.0%
Melting Point
116-120?C
Optical Rotation
|0 = -33? ?2? (C=1 in Dioxane)
Solution Test
2 mmol in 1 ml DCM + 1 ml DMF

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text