1-Benzyloxycarbonyl-4-oxo-L-proline, 97%
1-Benzyloxycarbonyl-4-oxo-L-proline, 97%
1-Benzyloxycarbonyl-4-oxo-L-proline, 97%
Thermo Scientific Chemicals

1-Benzyloxycarbonyl-4-oxo-L-proline, 97%

CAS: 64187-47-9 | C13H13NO5 | 263.25 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 H66054.03
亦稱為 H66054-03
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
規格
CAS64187-47-9
Chemical Name or Material1-Benzyloxycarbonyl-4-oxo-L-proline
ColorBrown to White
Formula Weight263.24
Health Hazard 1H315-H319-H335
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only