2-Amino-6-nitrobenzoic acid, 97%
2-Amino-6-nitrobenzoic acid, 97%
2-Amino-6-nitrobenzoic acid, 97%
Thermo Scientific Chemicals

2-Amino-6-nitrobenzoic acid, 97%

CAS: 50573-74-5 | C7H6N2O4 | 182.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66090.03
also known as H66090-03
Price (EUR)
42,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
42,30
Each
Chemical Identifiers
CAS50573-74-5
IUPAC Name2-amino-6-nitrobenzoic acid
Molecular FormulaC7H6N2O4
InChI KeyGGKYLHNARFFORH-UHFFFAOYSA-N
SMILESNC1=CC=CC(=C1C(O)=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only