Thermo Scientific Chemicals

4-Chloroindole, 97%, Thermo Scientific Chemicals

Catalog number: H66153.06
5 g, Each
Thermo Scientific Chemicals

4-Chloroindole, 97%, Thermo Scientific Chemicals

Catalog number: H66153.06
5 g, Each
Quantity
Catalog number: H66153.06
also known as H66153-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
25235-85-2
IUPAC Name
4-chloro-1H-indole
Molecular Formula
C8H6ClN
InChI Key
SVLZRCRXNHITBY-UHFFFAOYSA-N
SMILES
ClC1=C2C=CNC2=CC=C1
Form
Liquid.
Assay from Supplier's CofA
≥96.0%
Appearance (Color)
Pale yellow to pale brown
Proton NMR
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text