1-(Cyclopropylcarbonyl)piperazine hydrochloride, 97%
1-(Cyclopropylcarbonyl)piperazine hydrochloride, 97%
1-(Cyclopropylcarbonyl)piperazine hydrochloride, 97%
Thermo Scientific Chemicals

1-(Cyclopropylcarbonyl)piperazine hydrochloride, 97%

CAS: 1021298-67-8 | C8H15ClN2O | 190.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H66242.03
also known as H66242-03
Price (EUR)
73,60
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
73,60
Each
Specifications
Beilstein18312533
CAS1021298-67-8
Chemical Name or Material1-(Cyclopropylcarbonyl)piperazine hydrochloride
ColorWhite
Formula Weight190.67
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only