2-Amino-6-methoxypyridine, 97%
2-Amino-6-methoxypyridine, 97%
2-Amino-6-methoxypyridine, 97%
2-Amino-6-methoxypyridine, 97%
Thermo Scientific Chemicals

2-Amino-6-methoxypyridine, 97%

CAS: 17920-35-3 | C6H8N2O | 124.143 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 H66436.14
亦稱為 H66436-14
價格 (TWD)
-
申請報價
Quantity:
25 g
Request bulk or custom format
化學識別
CAS17920-35-3
IUPAC Name6-methoxypyridin-2-amine
Molecular FormulaC6H8N2O
InChI KeyDEUALFRBMNMGDS-UHFFFAOYSA-N
SMILESCOC1=CC=CC(N)=N1
檢視更多
規格Specification Sheet規格表
Assay (HPLC)>96.0%
It's an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It′s an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. William W. Paudler; David J. Pokorny; and James J. Good. Perchloroimidazo[1,2-a] pyridine. Synthesis and nucleophilic substitutions. Journal of Heterocyclic Chemistry.19718(1), 37-40.
  2. Fengrui Lang; Daniel Zewge; Ioannis N Houpis; R.P Volante. Amination of aryl halides using copper catalysis. Tetrahedron Letters.200142(19), 3251-3254.