5-Amino-1-methylbenzimidazole, 97%
5-Amino-1-methylbenzimidazole, 97%
5-Amino-1-methylbenzimidazole, 97%
Thermo Scientific Chemicals

5-Amino-1-methylbenzimidazole, 97%

CAS: 10394-38-4 | C8H9N3 | 147.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H66465.03
also known as H66465-03
Price (USD)
156.65
Online Exclusive
174.00
Save 17.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
156.65
Online Exclusive
174.00
Save 17.35 (10%)
Each
Specifications
Beilstein128912
CAS10394-38-4
Chemical Name or Material5-Amino-1-methylbenzimidazole
Formula Weight147.08
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only