6-Chloro-2-fluoronicotinic acid, 95%
6-Chloro-2-fluoronicotinic acid, 95%
6-Chloro-2-fluoronicotinic acid, 95%
Thermo Scientific Chemicals

6-Chloro-2-fluoronicotinic acid, 95%

CAS: 1211578-46-9 | C6H3ClFNO2 | 175.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H66484.03
also known as H66484-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1211578-46-9
IUPAC Name6-chloro-2-fluoropyridine-3-carboxylic acid
Molecular FormulaC6H3ClFNO2
InChI KeyGSSVAHBRTISODE-UHFFFAOYSA-N
SMILESOC(=O)C1=C(F)N=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.40%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only