ITIC-Cl, Thermo Scientific Chemicals
ITIC-Cl, Thermo Scientific Chemicals
ITIC-Cl, Thermo Scientific Chemicals
Thermo Scientific Chemicals

ITIC-Cl, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
250 mg
500 mg
Catalog number H66521.MC
also known as H66521-MC
Price (USD)
482.65
Special offer
Online exclusive
Ends: 30-Jun-2026
568.00
Save 85.35 (15%)
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
482.65
Special offer
Online exclusive
Ends: 30-Jun-2026
568.00
Save 85.35 (15%)
Each
Chemical Identifiers
CAS2253663-81-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
FormFlake or threadlike
Bandgap energy1.46 ? 0.02 eV
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only