3-Bromotetrahydro-4H-pyran-4-one, 90%, Thermo Scientific Chemicals
3-Bromotetrahydro-4H-pyran-4-one, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromotetrahydro-4H-pyran-4-one, 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H66595.MD
also known as H66595-MD
Price (USD)/ Each
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
IUPAC Name3-bromooxan-4-one
Molecular FormulaC5H7BrO2
InChI KeyZGULSQDBOSMIBZ-UHFFFAOYNA-N
SMILESBrC1COCCC1=O
Molecular Weight (g/mol)179.01
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)91.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only