Thermo Scientific Chemicals

N-Fmoc-D-glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H66624.06
5 g, Each
Thermo Scientific Chemicals

N-Fmoc-D-glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals

Catalog number: H66624.06
5 g, Each
Quantity
Catalog number: H66624.06
also known as H66624-06
Price (USD)
Price: 70.00
Online price: 59.65
Your price:
Quantity
-

Chemical Identifiers

CAS
104091-08-9
Assay (HPLC)
>97.5%

Description

It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text