N-Acetyl-L-isoleucine, 98%
N-Acetyl-L-isoleucine, 98%
N-Acetyl-L-isoleucine, 98%
Thermo Scientific Chemicals

N-Acetyl-L-isoleucine, 98%

CAS: 3077-46-1 | C8H15NO3 | 173.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H66771.14
also known as H66771-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS3077-46-1
IUPAC Name(2S,3S)-2-acetamido-3-methylpentanoic acid
Molecular FormulaC8H15NO3
InChI KeyJDTWZSUNGHMMJM-IFSCUNPJNA-N
SMILESCC[C@H](C)[C@H](NC(C)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only