Thermo Scientific Chemicals

N-Fmoc-L-valine N-succinimidyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H66946.14
25 g, Each
Thermo Scientific Chemicals

N-Fmoc-L-valine N-succinimidyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H66946.14
25 g, Each
Quantity
Catalog number: H66946.14
also known as H66946-14
Price (USD)
Price: 241.00
Online price: 207.65
Your price:
Quantity
-

Chemical Identifiers

CAS
130878-68-1
IUPAC Name
2,5-dioxopyrrolidin-1-yl (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylbutanoate
Molecular Formula
C24H24N2O6
InChI Key
JPJMNCROLRPFHI-QFIPXVFZSA-N
SMILES
CC(C)[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)ON1C(=O)CCC1=O
Assay (HPLC)
>94.0%

Description

N-Fmoc-L-valine N-succinimidyl ester is useful for solid phase peptide synthesis techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-valine N-succinimidyl ester is useful for solid phase peptide synthesis techniques.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text