3,4-Diaminobenzonitrile, 97%
3,4-Diaminobenzonitrile, 97%
3,4-Diaminobenzonitrile, 97%
Thermo Scientific Chemicals

3,4-Diaminobenzonitrile, 97%

CAS: 17626-40-3 | C7H7N3 | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H66954.14
also known as H66954-14
Price (EUR)
414,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
414,00
Each
Specifications
CAS17626-40-3
Chemical Name or Material3,4-Diaminobenzonitrile
ColorBrown
DOT InformationTransport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Formula Weight133.15
View more
3,4-Diaminobenzonitrile is usually used like synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Diaminobenzonitrile is usually used like synthetic intermediate.

Solubility
Soluble in dichloromethane, methanol and ethyl acetate.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. F. F. Stephens; and J. D. Bower. 348. The preparation of benziminazoles and benzoxazoles from Schiff's bases. Part II. J. Chem. Soc.19501722-1726.
  2. D. K. Geiger; and D. E. Parsons. 348. 3,4-Diamino­benzonitrile. Acta Cryst..2013E69, o452.