1-Propylphosphonic acid cyclic anhydride, 50+% w/w soln. in acetonitrile, Thermo Scientific Chemicals
1-Propylphosphonic acid cyclic anhydride, 50+% w/w soln. in acetonitrile, Thermo Scientific Chemicals
1-Propylphosphonic acid cyclic anhydride, 50+% w/w soln. in acetonitrile, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Propylphosphonic acid cyclic anhydride, 50+% w/w soln. in acetonitrile, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H66967.18
Price (EUR)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS68957-94-8
IUPAC Nametripropyl-1,3,5,2λ⁵,4λ⁵,6λ⁵-trioxatriphosphinane-2,4,6-trione
Molecular FormulaC9H21O6P3
InChI KeyPAQZWJGSJMLPMG-UHFFFAOYSA-N
SMILESCCCP1(=O)OP(=O)(CCC)OP(=O)(CCC)O1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Colorless to brown
Impurity content≤0.5 %[m/m] Propanphosphonic acid
Assay (unspecified)≥40.0 %[m/m] 1-Propylphosphonic acid
Impurity content≤50.0 %[m/m] Acetonitrile (solvent)
View more
1-Propylphosphonic acid cyclic anhydride is used as a reagent for amide/peptide bond formation. It is utilized for condensation reactions and alcohol oxidations. It is also used to promote the synthesis of hydroxamic acids from carboxylic acids. Further, it is employed as an efficient promoter for the Lossen rearrangement to synthesis urea and carbamate derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Propylphosphonic acid cyclic anhydride is used as a reagent for amide/peptide bond formation. It is utilized for condensation reactions and alcohol oxidations. It is also used to promote the synthesis of hydroxamic acids from carboxylic acids. Further, it is employed as an efficient promoter for the Lossen rearrangement to synthesis urea and carbamate derivatives.

Solubility
Miscible with dioxane, terahydrofuran, dimethyl formamide, polar and aprotic organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Madhu, C.; Basavaprabhu.; Vishwanatha, T. M.; Sureshbabu, V. V. T3P (propylphosphonic anhydride) mediated conversion of Nα-protected amino/peptide acids into thioacids. Tetrahedron Lett. 2012, 53 (11), 1406-1409.
  2. Nagendra, G.; Madhu, C.; Vishwanatha, T. M.; Sureshbabu, V. V. An expedient route for the reduction of carboxylic acids to alcohols employing 1-propanephosphonic acid cyclic anhydride as acid activator. Tetrahedron Lett. 2012, 53 (38), 5059-5063.
  3. Prasanna, T. S. R.; Raju, K. M. Quick Access to Bis(indolyl)methanes: T3P as a Novel Catalyst System. J. Korean Chem. Soc. 2012, 56 (1), 1406-1409.