Provide faster blots with higher efficiency using Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems.
Have Questions?
Change view
Catalog Number
Dimensions (L x W x H)
Capacity
HEP-3
50.4 x 39.7 x 5.1 cm
Transfer area: 35 x 45 cm
HEP-1
25 x 25 x 5 cm
Transfer area: 20 x 20 cm
2 Options
Catalog number HEP-3
Price (USD)
-
Dimensions (L x W x H):
50.4 x 39.7 x 5.1 cm
Capacity:
Transfer area: 35 x 45 cm
Provide faster blots with higher efficiency using Thermo Scientific™ Owl™ HEP Series Semidry Electroblotting Systems. Transfers can be done in as little as 15 minutes.
Fast, efficient transfer of proteins and nucleic acids to membranes
Buffer contained within blotting paper is sufficient for efficient transfer of molecules
Base contains stainless-steel cathode plate; lid has platinum/titanium anode plate