Search
View additional product information for HERAFreeze™ HFU B Series -86°C Upright Ultra-Low Temperature Freezer, 13 cu. ft. (368.1L) capacity, holds 240 boxes, 230V/50Hz, 12A, stainless-steel interior - FAQs (HFU240BSSV)
1 product FAQs found