Human Microsomes, 20 Donors, High CYP2D6 Activity

Catalog number: HMMC2D

Gibco™  Related applications: Hepatocytes & Hepatic Models | Pharma & Biopharma | Target-Based ADME⁄Tox Assays

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Human liver microsomes pooled from 20 different individual donors. All donors are equally represented for a truer population sample. These microsomes have been fully characterized for major cytochrome P450 activities and select Phase II enzymes.

Total protein concentration: 20 mg/ml

We provide liver subcellular fractions from a variety of tox species, including human, nonhuman primates (Cynomolgus Monkey and Rhesus Monkey), dog (Beagle), rat (Sprague-Dawley), mouse (CD-1 and Balb-c), and trout (Oncorhynchus mykiss). Custom species are available upon request.

For research use only. Not intended for human or animal therapeutic or diagnostic use.

Related Link

Learn more about our microsomes and subcellular fractions
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Age: Adult
Donor Source: Pooled
Format: Tube(s)
Gender: Mixed Gender
Sub-Cellular Localization: Microsomes
Tissue: Liver
Concentration: 20 mg⁄ml
Form: Frozen
Product Line: Gibco®
Product Size: 0.5 mL
Shipping Condition: Dry Ice
Species: Human

Contents & storage

Store between -60°C and -85°C.

Documents

Manuals & protocols