Product Image
Thermo Scientific™
Oxoid™ Anaerobic Plate Carrier, Stainless Steel
The Oxoid Anaerobic Plate Carrier is for use with Thermo Scientific™ Oxoid 3.5L Anaerobic Jars, Part No. HP0011A, HP0031A.
Have Questions?
Catalog NumberQuantity
HP0026A1 Ea.
Catalog number HP0026A
Price (MXN)
-
Quantity:
1 Ea.
Oxoid Anaerobic Plate Carrier is for use with Thermo Scientific™ Oxoid 3.5L Anaerobic Jars, Part No. HP0011A, HP0031A.
Specifications
MaterialStainless Steel
Quantity1 Ea.
TypeAnaerobic Plate Carrier
Unit SizeEach