HepaRG™ Serum-free Induction Medium Supplement (5X)
HepaRG™ Serum-free Induction Medium Supplement (5x) is a specialized media supplement for use in culturing HepaRG™ cells. HepaRG™ Serum-free InductionRead more
Have Questions?
Catalog Number
Quantity
HPRG750
8 mL
Catalog number HPRG750
Price (MXN)
10,756.40
Each
Quantity:
8 mL
Price (MXN)
10,756.40
Each
HepaRG™ Serum-free Induction Medium Supplement (5x) is a specialized media supplement for use in culturing HepaRG™ cells. HepaRG™ Serum-free Induction Medium Supplement (5x) is specifically used for induction of metabolic activity in HepaRG™ cells in a serum free environment. • Easy media preparation, just thaw, mix, and use • Reproducible, and biologically relevant results with HepaRG™ cells
Media Preparation Working media is prepared by the addition of thawed HepaRG™ Serum-free Induction Medium Supplement (5x) to 500 ml of William’s Medium E (A1217601 or 12551032) and 5 ml of GlutaMAX™-I (35050061). The simple two-step process helps reduce contamination opportunities.
Simplified, Relevant and Reproducible Studies HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).
HepaRG™ is a trademark of BioPredic International.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeHepaRG Cells
Concentration5 X
For Use With (Application)Drug Metabolism
Product LineGibco
Purity or Quality GradeResearch Grade, Cell Culture Grade