Product Image
Gibco™

HepaRG™ Thaw, Plate & General Purpose Medium Supplement (5X)

HepaRG™ Thaw, Plate & General Purpose Medium Supplement (5X) is a specialized media supplement for use in culturing HepaRG™ cells.
Have Questions?
Catalog NumberQuantity
HPRG77069 mL
Catalog number HPRG770
Price (USD)
459.00
Each
Quantity:
69 mL
Price (USD)
459.00
Each

HepaRG™ Thaw, Plate, & General Purpose Medium Supplement (5x) is a specialized media supplement for use in culturing HepaRG™ cells. HepaRG™ Thaw, Plate, & General Purpose Medium Supplement (5x) is specifically used for the initial thawing and propagation of cryopreserved HepaRG™ cells.

  • Easy media preparation, just thaw, mix, and use
  • Reproducible, and biologically relevant results with HepaRG™ cells

Media Preparation

Working media is prepared by the addition of thawed HepaRG™ Thaw, Plate, & General Purpose Medium Supplement (5x) to 500 ml of William's Medium E (A1217601 or 12551032) and 5 ml of GlutaMAX™-I (35050061). The simple two-step process helps reduce contamination opportunities.

Simplified, Relevant and Reproducible Studies

HepaRG™ cells exhibit many characteristics of primary human hepatocytes and are essentially a single donor. These features allow users to obtain physiological relevant results for metabolism-based drug-drug interactions without the concern of donor variability and limited lot sizes that come with relying on donor tissue. (Note: The cells we provide are terminally differentiated).

HepaRG™ is a trademark of BioPredic International.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeHepaRG Cells
Concentration5X
FormatBottle
Product LineGibco
Purity or Quality GradeResearch Grade, Cell Culture Grade
Quantity69 mL
Serum LevelStandard Serum
Shipping ConditionDry Ice
Tested ForSterility Testing (Bacteria & Fungi)
FormFrozen
Product TypeHepatocyte Supplement
SterilitySterile
Without AdditivesNo Glutamine
pH6 to 8
Unit SizeEach
Contents & Storage
Supplement should be stored at -20°C