One Lambda™ HybriType™ HLA Plus ABO Typing Flex Kit 96
One Lambda™ HybriType™ HLA Plus ABO Typing Flex Kit 96
Enables sequencing of 19 loci—including classical and non-classical HLA genes, MIC loci, and the ABO gene for comprehensive genotyping—using the One Lambda™ HybriType™ HLA Plus ABO Typing Flex Kit – 96 in combination with TypeStream™ Visual Software and a hybrid capture targeted enrichment approach.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad de procesamiento
HYB18ABOFX88 enriquecimientos
HYB18ABOFX4 enriquecimientos
Número de catálogo HYB18ABOFX8
Precio (MXN)
-
Capacidad de procesamiento:
8 enriquecimientos
HybriType™ sets itself apart with its hybrid capture technology, enabling robust performance while simultaneously simplifying the workflow process. Sequence 19 loci, covering HLA (classical and non-classical), MIC genes plus the ABO gene from blood or buccal sample extracted DNA. Its exceptional design delivers consistent and reliable high-resolution results across all loci on day 2. All reagents necessary for genomic library preparation and hybrid capture enrichment are included with this high-resolution NGS genotyping assay.
Simplified workflow
  • Support DNA from whole blood or buccal swabs across a wide input range with ready-to-use reagents.
  • Minimize hands-on time through single-step fragmentation, repair, and adenylation, one-step barcoding, and simplified post-capture washes.
Flexible, modular genotyping
  • Choose between two targeted panels: one for 11 classical HLA loci and another for 7 non-classical MHC loci.
  • Easily expand testing to include ABO typing with the HybriType™ ABO Flex Panel – 4 Enrichments, or maintain workflow continuity using the HybriType™ HLA Plus 18 Loci Flex Kit – 4 Enrichments when enrichment reagents run low.
Quality results
  • Mitigate PCR amplification bias and produce consistent balanced reads using target specific capture probes inherent to the HybriType™ NGS strategy.
Improved data quality
  • Achieve uniform, well-balanced data with hybrid capture–based targeted enrichment, while UDI indexing minimizes barcode hopping and misassignment.
Expand Understanding
  • Advance insight into the relationship between allele balance and zygosity with reported copy number values.
Especificaciones
Tipo de productoHLA and ABO Typing Assay
Cantidad96 Tests
Estado normativoFor Research Use Only. Not for use in diagnostic procedures.
SoftwareTypeStream Visual
Tipo de sistemaIllumina MiSeq, MiniSeq and iSeq systems
TécnicaHybrid Capture
Capacidad de procesamiento8 enriquecimientos
FormatoKit
Sequencing TypeShort Read Sequencing
Unit SizeEach
Contenido y almacenamiento
  • Qty:1; Box 1: Contains 6 reagents for preparing genomic libraries of up to 96 samples
  • Qty:1; Box 2: Contains 3 reagents, including 2 types of beads and 1 buffer solution
  • Qty:2; Box 3: Contains 9 reagents designed for processing 4 pooled libraries, optimized for hybrid capture enrichment of 18 loci (11 classical and 7 non-classical)
  • Qty:2; 1 reagent containing an oligonucleotide probe mix for hybrid capture and enrichment of the ABO blood group gene
Box 1, Box 3 and the oligonucleotide probe mix can be safely stored at -20°C and Box 2 can be safely stored at 4°C