Conjugates prepared with the thiol-reactive azide, succinimidyl ester can be detected with an alkyne-containing molecule in a click chemistry reaction.Read more
Have Questions?
Catalog Number
Quantity
I10188
1 mg
Catalog number I10188
Price (USD)
332.65
Online exclusive
443.00
Save 110.35
Each
Quantity:
1 mg
Price (USD)
332.65
Online exclusive
443.00
Save 110.35
Each
Conjugates prepared with the thiol-reactive azide, succinimidyl ester can be detected with an alkyne-containing molecule in a click chemistry reaction. Click chemistry describes a class of chemical reactions that use bio-orthogonal or biologically unique moities to label and detect a molecule of interest using a two-step procedure. The two-step reaction procedure involves a copper-catalyzed triazole formation of an azide and an alkyne. Click reactions have several characteristics: the reaction between the detection moieties is efficient; no extreme temperatures or solvents are required; the reaction product is stable; the components of the reaction are bioinert; and perhaps most importantly, no side reactions occur – the label and detection tags react selectively and specifically with one another. Unlike traditional chemical reactions utilizing succinimidyl esters or maleimides that target amines and sulfhydryls – functional groups that are not unique – click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityThiol
FormatSolid
Label or DyeAzide
Molecular Weight (g/mol)310.14 Da
Product TypeIodoacetamide Azide
Quantity1 mg
Reactive GroupIodoacetamide
Reactive MoietyAlkyl Halide, Iodoacetamide
Shipping ConditionRoom Temperature
SolubilityDMSO (Dimethylsulfoxide)
Label TypeClick Chemistry Label
Product LineMolecular Probes
Unit SizeEach
Contents & Storage
Store at ≤-20°C, desiccated and protected from light.