Image-IT™ LIVE Green Reactive Oxygen Species Detection Kit, for microscopy
Invitrogen™
Image-IT™ LIVE Green Reactive Oxygen Species Detection Kit, for microscopy
Image-iT™ LIVE Green Reactive Oxygen Species (ROS) Detection Kit는 살아있는 세포에서 ROS 검출에 필요한 주요 시약을 제공합니다. 이 분석은 생존자세히 알아보기
Have Questions?
카탈로그 번호수량
I360071 Kit
카탈로그 번호 I36007
제품 가격(KRW)
607,000
온라인 행사
Ends: 31-Dec-2026
693,000
할인액 86,000
Each
수량:
1 Kit
제품 가격(KRW)
607,000
온라인 행사
Ends: 31-Dec-2026
693,000
할인액 86,000
Each
Image-iT™ LIVE Green Reactive Oxygen Species (ROS) Detection Kit는 살아있는 세포에서 ROS 검출에 필요한 주요 시약을 제공합니다. 이 분석은 생존 세포에서 신뢰할 수 있는 ROS 형광표지자인 5-(and-6)-carboxy-2',7'-dichlorodihydrofluorescein diacetate (carboxy-H2DCFDA)를 기반으로 합니다. Carboxy-H2DCFDA에 추가하여 이 키트는 양성 대조로 ROS 생성 tert-butyl hydroperoxide (TBHP) 공통 유도자와 청색 형광 세포 투과성 핵산 염색 Hoechst 33342를 제공합니다.
For Research Use Only. Not for use in diagnostic procedures.
사양
수량1 Kit
제품라인Image-iT
제품 유형ROS Detection Kit
Unit SizeEach
인용 및 참조 문헌 (34)
인용 및 참조 문헌
Abstract
Immunoglobulin light chains generate hydrogen peroxide.
Authors:Wang PX, Sanders PW,
Journal:J Am Soc Nephrol
PubMed ID:17360948
'As low molecular weight proteins, restriction from glomerular filtration is minimized, permitting significant amounts of Ig light chains to be endocytosed into the proximal tubule epithelium, particularly in plasma cell dyscrasias. Recent studies have shown that this effect of concentrating light chains within the proximal tubule alters cell function. This ... More
Estrogen prevents cardiomyocyte apoptosis through inhibition of reactive oxygen species and differential regulation of p38 kinase isoforms.
Authors:Kim JK, Pedram A, Razandi M, Levin ER
Journal:J Biol Chem
PubMed ID:16407188
'From human and animal studies, estrogen is known to protect the myocardium from an ischemic insult. However, there is limited knowledge regarding mechanisms by which estrogen directly protects cardiomyocytes. In this report, we employed an in vitro model, in which cultured rat cardiomyocytes underwent prolonged hypoxia followed by reoxygenation (H/R), ... More
24-dehydrocholesterol reductase/seladin-1: a key protein differentially involved in adrenocorticotropin effects observed in human and rat adrenal cortex.
Authors:Battista MC, Roberge C, Martinez A, Gallo-Payet N,
Journal:Endocrinology
PubMed ID:19520779
'DHCR24 (24-dehydrocholesterol reductase), or seladin-1, is one of the most expressed genes in the adrenal gland. Because the rat and human adult adrenal cortex differ in their respective functional properties, the aim of the present study was to verify whether seladin-1 may be differentially involved in basal and ACTH-stimulated steroidogenesis ... More
Tubacin kills Epstein-Barr virus (EBV)-Burkitt lymphoma cells by inducing reactive oxygen species and EBV lymphoblastoid cells by inducing apoptosis.
Authors:Kawada J, Zou P, Mazitschek R, Bradner JE, Cohen JI,
Journal:J Biol Chem
PubMed ID:19386607
'Tubacin is a small molecule inhibitor of histone deacetylase 6 and blocks aggresome activity. We found that Epstein-Barr virus (EBV)-positive Burkitt lymphoma (BL) cells were generally killed by lower doses of tubacin than EBV-transformed lymphoblastoid cells (LCLs) or EBV-negative BL cells. Tubacin induced apoptosis of LCLs, which was inhibited by ... More
HER-2 signaling, acquisition of growth factor independence, and regulation of biological networks associated with cell transformation.
Authors:Bollig-Fischer A, Dziubinski M, Boyer A, Haddad R, Giroux CN, Ethier SP,
Journal:Cancer Res
PubMed ID:20736364
'Activated oncogenes are the dominant drivers of malignant progression in human cancer, yet little is known about how the transformation from proto-oncogene to activated oncogene drives the expression of transformed phenotypes. An isogenic model of HER-2-mediated transformation of human mammary epithelial cells was used along with HER-2-amplified human breast cancers ... More